missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (phenylthio)acetate, 97%, Thermo Scientific™

Product Code. 11482948
Click to view available options
Quantity:
2.5 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7605-25-6
Molecular Formula C10H12O2S
Molecular Weight (g/mol) 196.264
MDL Number MFCD00145107
InChI Key SEDRTXNDGKRHBL-UHFFFAOYSA-N
Synonym ethyl phenylthio acetate, ethyl 2-phenylthio acetate, ethyl 2-phenylsulfanyl acetate, acetic acid, phenylthio-, ethyl ester, ethyl 2-phenylthioacetate, phenylthio acetic acid ethyl ester, phenylthio acetic acid, ethyl ester, ethyl phenylsulfanyl acetate, pubchem10853
PubChem CID 97312
IUPAC Name ethyl 2-phenylsulfanylacetate
SMILES CCOC(=O)CSC1=CC=CC=C1

Specifications

Boiling Point 143°C to 145°C (14 mmHg)
Quantity 2.5 g
Formula Weight 196.27
Percent Purity 97%
Chemical Name or Material Ethyl (phenylthio)acetate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'