missing translation for 'onlineSavingsMsg'
Learn More

Ethyl potassium malonate, 98%, Thermo Scientific Chemicals

Product Code. 10162951
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6148-64-7
Molecular Formula C5H7KO4
Molecular Weight (g/mol) 170.2
MDL Number MFCD00035603
InChI Key WVUCPRGADMCTBN-UHFFFAOYSA-M
Synonym ethyl potassium malonate, potassium 3-ethoxy-3-oxopropanoate, potassium ethyl malonate, potassium monoethyl malonate, monoethyl malonate potassium salt, malonic acid monoethyl ester potassium salt, monoethyl potassium malonate, ethyl malonate potassium salt, propanedioic acid, monoethyl ester, potassium salt, ethylpotassiummalonate
PubChem CID 3446434
IUPAC Name potassium;3-ethoxy-3-oxopropanoate
SMILES CCOC(=O)CC(=O)[O-].[K+]

Specifications

Melting Point 200.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula KOOCCH2COOC2H5
Quantity 25 g
Beilstein 02,III,1609
Fieser 15,156
Solubility Information Solubility in water: 2.53g/L (25°C)
Formula Weight 170.2
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material Ethyl potassium malonate, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl potassium malonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'