Learn More
Ethyl propionate, 99+%, Thermo Scientific Chemicals
£14.10 - £63.40
Chemical Identifiers
CAS | 105-37-3 |
---|---|
Molecular Formula | C5H10O2 |
Molecular Weight (g/mol) | 102.13 |
MDL Number | MFCD00009308 |
InChI Key | FKRCODPIKNYEAC-UHFFFAOYSA-N |
Synonym | ethyl propionate, propanoic acid, ethyl ester, propionic ester, propionic ether, propionic acid, ethyl ester, ethylpropionate, propionate d'ethyle, propanoic acid ethyl ester, ethyl n-propionate, propionic acid ethyl ester |
PubChem CID | 7749 |
ChEBI | CHEBI:41330 |
IUPAC Name | ethyl propanoate |
SMILES | CCC(=O)OCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10367510
|
Thermo Scientific Acros
150500250 |
25 mL | Glass bottle |
£14.10
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10183050
|
Thermo Scientific Acros
150505000 |
500 mL | Glass bottle |
£21.90
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10285290
|
Thermo Scientific Acros
150500010 |
1 L | Glass bottle |
£36.60
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10279503
|
Thermo Scientific Acros
150500025 |
2.5 L | Glass bottle |
£63.40
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
105-37-3 | |
102.13 | |
FKRCODPIKNYEAC-UHFFFAOYSA-N | |
7749 | |
ethyl propanoate |
C5H10O2 | |
MFCD00009308 | |
ethyl propionate, propanoic acid, ethyl ester, propionic ester, propionic ether, propionic acid, ethyl ester, ethylpropionate, propionate d'ethyle, propanoic acid ethyl ester, ethyl n-propionate, propionic acid ethyl ester | |
CHEBI:41330 | |
CCC(=O)OCC |
Specifications
1mg KOH/g max. | |
-73°C | |
0.8870g/mL | |
12°C | |
99% min. (GC) | |
C2H5CO2C2H5 | |
MFCD00009308 | |
01,385 | |
0.887 | |
Solubility in water: 25g/L water (15°C). Other solubilities: miscible with alcohol,ether and propylene glycol | |
CCC(=O)OCC | |
102.13 | |
CHEBI:41330 | |
102.13 | |
Liquid |
105-37-3 | |
Colorless to Yellow | |
99°C | |
Authentic | |
C5H10O2 | |
1.3830 to 1.387 | |
02, 240 | |
15, 3899 | |
ethyl propionate, propanoic acid, ethyl ester, propionic ester, propionic ether, propionic acid, ethyl ester, ethylpropionate, propionate d'ethyle, propanoic acid ethyl ester, ethyl n-propionate, propionic acid ethyl ester | |
FKRCODPIKNYEAC-UHFFFAOYSA-N | |
ethyl propanoate | |
7749 | |
<1 mPa.s (20°C) | |
99+% | |
Ethyl propionate |
Safety and Handling
GHS H Statement:
Highly flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-291-4
RUO – Research Use Only