missing translation for 'onlineSavingsMsg'
Learn More

Ethyl pyrazolo[1,5-a]pyrimidine-3-carboxylate, 95%, Thermo Scientific™

Product Code. 15428725
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115932-00-8
Molecular Formula C9H9N3O2
Molecular Weight (g/mol) 191.19
MDL Number MFCD00847745
InChI Key JEMAMNBFHPIPCR-UHFFFAOYSA-N
Synonym ethyl pyrazolo 1,5-a pyrimidine-3-carboxylate, pyrazolo 1,5-a pyrimidine-3-carboxylic acid ethyl ester, pyrazolo 1,5-a pyrimidine-3-carboxylic acid, ethyl ester, 3-ethoxycarbonyl pyrazolo 1,5-a pyrimidine, ethylpyrazolo 1,5-a pyrimidine-3-carboxylate, ethyl pyrazolo 1,5-a pyrimidine-3-carboxylate,, pubchem20847, ethyl pyrazolo 1,5-a pyrim-idine-3-carboxylate, ethyl 8-hydropyrazolo 1,5-a pyrimidine-3-carboxylate
PubChem CID 7021267
IUPAC Name ethyl pyrazolo[1,5-a]pyrimidine-3-carboxylate
SMILES CCOC(=O)C1=C2N=CC=CN2N=C1

Specifications

Melting Point 210°C to 212°C
Quantity 250 mg
Formula Weight 191.19
Percent Purity 95%
Chemical Name or Material Ethyl pyrazolo[1,5-a]pyrimidine-3-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'