missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl pyrazolo[1,5-a]pyrimidine-3-carboxylate, 95%, Thermo Scientific™
£48.60 - £133.00
Chemical Identifiers
CAS | 115932-00-8 |
---|---|
Molecular Formula | C9H9N3O2 |
Molecular Weight (g/mol) | 191.19 |
MDL Number | MFCD00847745 |
InChI Key | JEMAMNBFHPIPCR-UHFFFAOYSA-N |
Synonym | ethyl pyrazolo 1,5-a pyrimidine-3-carboxylate, pyrazolo 1,5-a pyrimidine-3-carboxylic acid ethyl ester, pyrazolo 1,5-a pyrimidine-3-carboxylic acid, ethyl ester, 3-ethoxycarbonyl pyrazolo 1,5-a pyrimidine, ethylpyrazolo 1,5-a pyrimidine-3-carboxylate, ethyl pyrazolo 1,5-a pyrimidine-3-carboxylate,, pubchem20847, ethyl pyrazolo 1,5-a pyrim-idine-3-carboxylate, ethyl 8-hydropyrazolo 1,5-a pyrimidine-3-carboxylate |
PubChem CID | 7021267 |
IUPAC Name | ethyl pyrazolo[1,5-a]pyrimidine-3-carboxylate |
SMILES | CCOC(=O)C1=C2N=CC=CN2N=C1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
115932-00-8 | |
191.19 | |
JEMAMNBFHPIPCR-UHFFFAOYSA-N | |
7021267 | |
CCOC(=O)C1=C2N=CC=CN2N=C1 |
C9H9N3O2 | |
MFCD00847745 | |
ethyl pyrazolo 1,5-a pyrimidine-3-carboxylate, pyrazolo 1,5-a pyrimidine-3-carboxylic acid ethyl ester, pyrazolo 1,5-a pyrimidine-3-carboxylic acid, ethyl ester, 3-ethoxycarbonyl pyrazolo 1,5-a pyrimidine, ethylpyrazolo 1,5-a pyrimidine-3-carboxylate, ethyl pyrazolo 1,5-a pyrimidine-3-carboxylate,, pubchem20847, ethyl pyrazolo 1,5-a pyrim-idine-3-carboxylate, ethyl 8-hydropyrazolo 1,5-a pyrimidine-3-carboxylate | |
ethyl pyrazolo[1,5-a]pyrimidine-3-carboxylate |
Specifications
115932-00-8 | |
C9H9N3O2 | |
ethyl pyrazolo 1,5-a pyrimidine-3-carboxylate, pyrazolo 1,5-a pyrimidine-3-carboxylic acid ethyl ester, pyrazolo 1,5-a pyrimidine-3-carboxylic acid, ethyl ester, 3-ethoxycarbonyl pyrazolo 1,5-a pyrimidine, ethylpyrazolo 1,5-a pyrimidine-3-carboxylate, ethyl pyrazolo 1,5-a pyrimidine-3-carboxylate,, pubchem20847, ethyl pyrazolo 1,5-a pyrim-idine-3-carboxylate, ethyl 8-hydropyrazolo 1,5-a pyrimidine-3-carboxylate | |
CCOC(=O)C1=C2N=CC=CN2N=C1 | |
191.19 | |
191.19 | |
Ethyl pyrazolo[1,5-a]pyrimidine-3-carboxylate |
210°C to 212°C | |
MFCD00847745 | |
JEMAMNBFHPIPCR-UHFFFAOYSA-N | |
ethyl pyrazolo[1,5-a]pyrimidine-3-carboxylate | |
7021267 | |
95% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures