missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl stearate, 97%, Thermo Scientific Chemicals
£24.50 - £58.20
Chemical Identifiers
CAS | 111-61-5 |
---|---|
Molecular Formula | C20H40O2 |
Molecular Weight (g/mol) | 312.52 |
MDL Number | MFCD00009006 |
InChI Key | MVLVMROFTAUDAG-UHFFFAOYSA-N |
Synonym | ethyl stearate, stearic acid ethyl ester, octadecanoic acid, ethyl ester, ethyl n-octadecanoate, stearic acid, ethyl ester, dicyclohexylammonium sulfate, unii-c64rtc734w, fema no. 3490, ethyl-stearate, ethyloctadecanoate |
PubChem CID | 8122 |
ChEBI | CHEBI:84936 |
IUPAC Name | ethyl octadecanoate |
SMILES | CCCCCCCCCCCCCCCCCC(=O)OCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10092620
|
Thermo Scientific Acros
207480050 |
5 g | Glass bottle |
£24.50
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10378150
|
Thermo Scientific Acros
207480250 |
25 g | Glass bottle |
£58.20
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
111-61-5 | |
312.52 | |
MVLVMROFTAUDAG-UHFFFAOYSA-N | |
8122 | |
ethyl octadecanoate |
C20H40O2 | |
MFCD00009006 | |
ethyl stearate, stearic acid ethyl ester, octadecanoic acid, ethyl ester, ethyl n-octadecanoate, stearic acid, ethyl ester, dicyclohexylammonium sulfate, unii-c64rtc734w, fema no. 3490, ethyl-stearate, ethyloctadecanoate | |
CHEBI:84936 | |
CCCCCCCCCCCCCCCCCC(=O)OCC |
Specifications
111-61-5 | |
White | |
>112°C | |
96% min. (GC) | |
CH3(CH2)16CO2C2H5 | |
02, 379 | |
ethyl stearate, stearic acid ethyl ester, octadecanoic acid, ethyl ester, ethyl n-octadecanoate, stearic acid, ethyl ester, dicyclohexylammonium sulfate, unii-c64rtc734w, fema no. 3490, ethyl-stearate, ethyloctadecanoate | |
MVLVMROFTAUDAG-UHFFFAOYSA-N | |
ethyl octadecanoate | |
8122 | |
312.52 | |
Crystalline Mass |
30°C to 39°C | |
213°C to 215°C (15.0 mmHg) | |
Authentic | |
C20H40O2 | |
MFCD00009006 | |
15, 8930 | |
Solubility in water: insoluble in water. Other solubilities: soluble in alcohol,ether,acetone | |
CCCCCCCCCCCCCCCCCC(=O)OCC | |
312.52 | |
CHEBI:84936 | |
97% | |
Ethyl stearate, 97% |
Safety and Handling
missing translation for 'einecsNumber' : 203-887-4
missing translation for 'rtecsNumber' : WI3600000
missing translation for 'tsca' : TSCA
RUO – Research Use Only