missing translation for 'onlineSavingsMsg'
Learn More

Ethyl succinyl chloride, 95%, Thermo Scientific Chemicals

Product Code. 10489670
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14794-31-1
Molecular Formula C6H9ClO3
Molecular Weight (g/mol) 164.59
MDL Number MFCD00000751
InChI Key IXZFDJXHLQQSGQ-UHFFFAOYSA-N
Synonym ethyl succinyl chloride, ethyl 3-chloroformyl propionate, ethyl 4-chloro-4-oxobutyrate, butanoic acid, 4-chloro-4-oxo-, ethyl ester, 3-carboethoxypropionyl chloride, ethoxycarbonylpropionyl chloride, 3-ethoxycarbonylpropionyl chloride, propionic acid, 3-chloroformyl-, ethyl ester, 4-chloro-4-oxo-butanoic acid ethyl ester, beta carbethoxy propionyl chloride
PubChem CID 84648
IUPAC Name ethyl 4-chloro-4-oxobutanoate
SMILES CCOC(=O)CCC(=O)Cl

Specifications

Color Colorless to Yellow
Density 1.1575g/mL
Boiling Point 88.0°C to 90.0°C (11.0 mmHg)
Flash Point 84°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula ClCOCH2CH2CO2C2H5
Refractive Index 1.4350 to 1.4390
Quantity 25 g
Beilstein 02, 613
Specific Gravity 1.1575
Formula Weight 164.59
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Ethyl succinyl chloride, 95%
Show More Show Less
compliance-icons
Product Identifier
  • Ethyl succinyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'