Learn More
Ethyl succinyl chloride, 95%, Thermo Scientific Chemicals
£27.90 - £320.00
Chemical Identifiers
CAS | 14794-31-1 |
---|---|
Molecular Formula | C6H9ClO3 |
Molecular Weight (g/mol) | 164.585 |
MDL Number | MFCD00000751 |
InChI Key | IXZFDJXHLQQSGQ-UHFFFAOYSA-N |
Synonym | ethyl succinyl chloride, ethyl 3-chloroformyl propionate, ethyl 4-chloro-4-oxobutyrate, butanoic acid, 4-chloro-4-oxo-, ethyl ester, 3-carboethoxypropionyl chloride, ethoxycarbonylpropionyl chloride, 3-ethoxycarbonylpropionyl chloride, propionic acid, 3-chloroformyl-, ethyl ester, 4-chloro-4-oxo-butanoic acid ethyl ester, beta carbethoxy propionyl chloride |
PubChem CID | 84648 |
IUPAC Name | ethyl 4-chloro-4-oxobutanoate |
SMILES | CCOC(=O)CCC(=O)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11460683
|
Thermo Scientific Alfa Aesar
A12734.14 |
25 g |
£27.90
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11470683
|
Thermo Scientific Alfa Aesar
A12734.22 |
100 g |
£81.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11480683
|
Thermo Scientific Alfa Aesar
A12734.36 |
500 g |
£320.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14794-31-1 | |
164.585 | |
IXZFDJXHLQQSGQ-UHFFFAOYSA-N | |
84648 | |
CCOC(=O)CCC(=O)Cl |
C6H9ClO3 | |
MFCD00000751 | |
ethyl succinyl chloride, ethyl 3-chloroformyl propionate, ethyl 4-chloro-4-oxobutyrate, butanoic acid, 4-chloro-4-oxo-, ethyl ester, 3-carboethoxypropionyl chloride, ethoxycarbonylpropionyl chloride, 3-ethoxycarbonylpropionyl chloride, propionic acid, 3-chloroformyl-, ethyl ester, 4-chloro-4-oxo-butanoic acid ethyl ester, beta carbethoxy propionyl chloride | |
ethyl 4-chloro-4-oxobutanoate |
Specifications
14794-31-1 | |
88°C to 90°C (11 mmHg) | |
Acrid | |
1.437 | |
UN3265 | |
Moisture sensitive | |
IXZFDJXHLQQSGQ-UHFFFAOYSA-N | |
ethyl 4-chloro-4-oxobutanoate | |
84648 | |
97% |
1.163 | |
84°C (183°F) | |
C6H9ClO3 | |
MFCD00000751 | |
1766126 | |
ethyl succinyl chloride, ethyl 3-chloroformyl propionate, ethyl 4-chloro-4-oxobutyrate, butanoic acid, 4-chloro-4-oxo-, ethyl ester, 3-carboethoxypropionyl chloride, ethoxycarbonylpropionyl chloride, 3-ethoxycarbonylpropionyl chloride, propionic acid, 3-chloroformyl-, ethyl ester, 4-chloro-4-oxo-butanoic acid ethyl ester, beta carbethoxy propionyl chloride | |
CCOC(=O)CCC(=O)Cl | |
164.585 | |
164.59 | |
Ethyl succinyl chloride |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H227-H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 238-855-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only