Learn More
Masterflex™ Ethyl Vinyl Acetate Microbore Tubing
Use this tubing for biological testing. Saint Gobain Performance Plastics™ Ethyl Vinyl Acetate Microbore Tubing is biologically stable, nontoxic and non-contaminating.
Brand:  Masterflex™ 95712-32
 View more versions of this product
Product Code. 11927885
  • £72.60 / 15m
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Translucent cross-linked tubing flexible micro-line tubing
  • Virtually 100% elastic memory, returns to its original shape with gentle heating
  • Resin is USP 23 Class VI and National Formulary Vol. 14 specifications
  • Sterilize with autoclave, ethylene oxide, gamma irradiation or dry heat
Specifications
Specifications
1.27 mm
3.23 mm
Solvent/Hydrocarbon Autoanalysis/Microbore color-coded stopped tubing
1 Pack
Translucent Yellow
EVA
Solvent/Hydrocarbon Autoanalysis/Microbore color-coded stopped tubing
-50°C
32
74°C