missing translation for 'onlineSavingsMsg'
Learn More
Ethylaluminium sesquichloride, 0.4M solution in hexane, AcroSeal™, Thermo Scientific Chemicals Product Code.: 10311803

Ethylaluminium sesquichloride, 0.4M solution in hexane, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10311803
800 mL, AcroSeal™ Glass Bottle
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10311803

Brand: Thermo Scientific Acros 185468000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12075-68-2, 110-54-3
Molecular Formula C6H15Al2Cl3
Molecular Weight (g/mol) 247.51
MDL Number MFCD00044852
InChI Key LDXMUMPEDOQULK-UHFFFAOYSA-K
Synonym ethylaluminum sesquichloride, ethylaluminium sesquichloride, sesquiethylaluminum chloride, easc, aluminum, trichlorotriethyldi, ethylaluminum sesquichioride, ethylaluminum sesquichloride solution, di-, i-chlorochlorotriethyldi-aluminum, chloro diethyl alumane; dichloro ethyl alumaneShow MoreShow Less
PubChem CID 25508
IUPAC Name chloro(diethyl)alumane;dichloro(ethyl)alumane
SMILES CC[Al](CC)Cl.CC[Al](Cl)Cl

Specifications

Color Colorless
Density 0.7010g/mL
Flash Point -22°C
Packaging AcroSeal™ Glass Bottle
Linear Formula (C2H5)2AlCl·Cl2AlC2H5
Quantity 800 mL
Specific Gravity 0.701
Solubility Information Solubility in water: vigorous reaction. Other solubilities: miscible with saturated aliphatic and aromatic,hydrocarbons
Formula Weight 247.51
Percent Purity 14.5 to 15.5% (on Al basis)
Physical Form Liquid
Chemical Name or Material Ethylaluminium sesquichloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0453960 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number185468000,185461000
Lot NumberA0453961 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number185468000,185461000
Lot NumberA0427631 Certificate TypeCertificate of Analysis Date27/02/2024 Catalog Number185468000,185461000
Lot NumberA0427630 Certificate TypeCertificate of Analysis Date27/02/2024 Catalog Number185468000,185461000
Lot NumberA0453960 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number185468000,185461000
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethylaluminium sesquichloride, 0.4M solution in hexane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H336-May cause drowsiness or dizziness.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethylaluminium sesquichloride, 0.4M solution in hexane, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.