missing translation for 'onlineSavingsMsg'
Learn More

Ethylbenzene, 99.8%, pure, Thermo Scientific Chemicals

Product Code. 10339444
Click to view available options
Quantity:
25 mL
500 mL
1 L
2.5 L
25 L
Packaging:
Glass bottle
Metal Drum
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-41-4
MDL Number MFCD00011647
InChI Key YNQLUTRBYVCPMQ-UHFFFAOYSA-N
Synonym phenylethane, ethylbenzol, benzene, ethyl, aethylbenzol, ethylenzene, ethyl benzene, ethylbenzeen, etilbenzene, etylobenzen, alpha-methyltoluene
PubChem CID 7500
ChEBI CHEBI:16101
IUPAC Name ethylbenzene
SMILES CCC1=CC=CC=C1

Specifications

Melting Point -95.0°C
Density 0.8600g/mL
Boiling Point 136.0°C
Flash Point 22°C
Infrared Spectrum Authentic
Assay Percent Range 99.7% min. (GC)
Packaging Metal Drum
Molecular Formula C8H10
Linear Formula C6H5C2H5
Refractive Index 1.4940 to 1.4970
Quantity 25 L
Beilstein 05, II, 274
Merck Index 15, 3820
Specific Gravity 0.86
Solubility Information Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in alcohol, benzene, carbon tetrachloride,, ether and sulfur dioxide, slightly soluble in chloroform, insoluble in ammonia
Molecular Weight (g/mol) 106.17
Viscosity 0.63 mPa.s (20°C)
Formula Weight 106.17
Percent Purity 99.8%
Grade Pure
Physical Form Liquid
Chemical Name or Material Ethylbenzene, 99.8%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethylbenzene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H332-Harmful if inhaled.
  • H304-May be fatal if swallowed and enters airways.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.