missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ethyle1-methylcyclopropane-1-carboxylate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
25 g
Specifications
Specifications
CAS Min % | 97.0 |
Density | 0.9180g/mL |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Linear Formula | CH3C3H4C(O)OCH2CH3 |
Quantity | 1 g |
Synonym | ethyl 1-methylcyclopropanecarboxylate, 1-methylcyclopropanecarboxylic acid ethyl ester, ethyl1-methylcyclopropanecarboxylate, cyclopropanecarboxylic acid, 1-methyl-, ethyl ester, cyclopropanecarboxylicacid, 1-methyl-, ethyl ester, ethyl 1-methylcyclopropane-carboxylate, 1-methylcyclopropanoic acid ethyl ester, 1-ethoxycarbonyl-1-methylcyclopropane, ethyl 1-methylcyclopropan-1-carboxylate, ethyl 1-methyl cyclopropane-1-carboxylate |
Formula Weight | 128.17 |
Percent Purity | 97% |
Chemical Name or Material | Ethyl 1-methylcyclopropane-1-carboxylate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ethyle1-methylcyclopropane-1-carboxylate, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction