missing translation for 'onlineSavingsMsg'
Learn More

ethyle2-bromo-4-methyl-1,3-thiazole-5-carboxylate, 97%, Thermo Scientific™

Product Code. 10703443
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 22900-83-0
Molecular Formula C7H8BrNO2S
Molecular Weight (g/mol) 250.11
MDL Number MFCD03791227
InChI Key CFBIOWPDDZPIDP-UHFFFAOYSA-N
Synonym ethyl 2-bromo-4-methylthiazole-5-carboxylate, 5-thiazolecarboxylic acid, 2-bromo-4-methyl-, ethyl ester, 2-bromo-4-methyl-thiazole-5-carboxylic acid ethyl ester, ethyl 2-bromo-4-methyl-5-thiazolecarboxylate, 2-bromo-4-methyl-5-thiazolecarboxylic acid ethyl ester, 2-bromo-4-methylthiazole-5-carboxylic acid ethyl ester, maybridge3_004543, ksc207i5t, ethyl2-bromo-4-methyl-1,3-thiazole-5-carboxylate, ethylbromomethylthiazolecarboxylate
PubChem CID 2824057
IUPAC Name ethyl 2-bromo-4-methyl-1,3-thiazole-5-carboxylate
SMILES CCOC(=O)C1=C(N=C(S1)Br)C

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 250.12
Percent Purity 97%
Chemical Name or Material ethyle2-bromo-4-methyl-1, 3-thiazole-5-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'