missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 2-methylnicotinate, 97%, Thermo Scientific™
£15.38 - £51.32
Chemical Identifiers
CAS | 1721-26-2 |
---|---|
Molecular Formula | C9H11NO2 |
Molecular Weight (g/mol) | 165.19 |
MDL Number | MFCD00006336 |
InChI Key | TZORNSWZUZDUCU-UHFFFAOYSA-N |
Synonym | ethyl 2-methylnicotinate, 2-methylnicotinic acid ethyl ester, 2-methyl-nicotinic acid ethyl ester, 3-pyridinecarboxylic acid, 2-methyl-, ethyl ester, ethyl 2-methyl nicotinate, ethyl 2-methyl-3-pyridinecarboxylate, 2-methyl-3-pyridinecarboxylic acid ethyl ester, 2-methylpyridine-3-carboxylic acid ethyl ester, ethyl 2-methylpyridine-3-carboxylate∼2-methylnicotinic acid ethyl ester, pubchem12727 |
PubChem CID | 74401 |
IUPAC Name | ethyl 2-methylpyridine-3-carboxylate |
SMILES | CCOC(=O)C1=CC=CN=C1C |
Description
Chemical Identifiers
1721-26-2 | |
165.19 | |
TZORNSWZUZDUCU-UHFFFAOYSA-N | |
74401 | |
CCOC(=O)C1=CC=CN=C1C |
C9H11NO2 | |
MFCD00006336 | |
ethyl 2-methylnicotinate, 2-methylnicotinic acid ethyl ester, 2-methyl-nicotinic acid ethyl ester, 3-pyridinecarboxylic acid, 2-methyl-, ethyl ester, ethyl 2-methyl nicotinate, ethyl 2-methyl-3-pyridinecarboxylate, 2-methyl-3-pyridinecarboxylic acid ethyl ester, 2-methylpyridine-3-carboxylic acid ethyl ester, ethyl 2-methylpyridine-3-carboxylate∼2-methylnicotinic acid ethyl ester, pubchem12727 | |
ethyl 2-methylpyridine-3-carboxylate |
Specifications
1721-26-2 | |
1.0700g/mL | |
103°C | |
96% min. (GC) | |
C9H11NO2 | |
1.07 | |
TZORNSWZUZDUCU-UHFFFAOYSA-N | |
ethyl 2-methylpyridine-3-carboxylate | |
74401 | |
97% | |
Ethyl 2-methylnicotinate |
Colorless to Orange | |
126°C to 127°C (24.0 mmHg) | |
Authentic | |
Glass Bottle | |
MFCD00006336 | |
ethyl 2-methylnicotinate, 2-methylnicotinic acid ethyl ester, 2-methyl-nicotinic acid ethyl ester, 3-pyridinecarboxylic acid, 2-methyl-, ethyl ester, ethyl 2-methyl nicotinate, ethyl 2-methyl-3-pyridinecarboxylate, 2-methyl-3-pyridinecarboxylic acid ethyl ester, 2-methylpyridine-3-carboxylic acid ethyl ester, ethyl 2-methylpyridine-3-carboxylate∼2-methylnicotinic acid ethyl ester, pubchem12727 | |
CCOC(=O)C1=CC=CN=C1C | |
165.19 | |
165.19 | |
Liquid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 217-013-4
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Ethyl 2-methylnicotinate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only