missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ethyle3-amino-4-(methylamino)benzoate, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | ethyl 3-amino-4-methylamino benzoate, 3-amino-4-methylamino benzoic acid ethyl ester, ethyl 4-methylamino-3-aminobenzoate, ethyl 3-azanyl-4-methylamino benzoate, ethyl3-amino-4-methylamino benzoate, 3-amino-4-methylaminobenzoic acid ethyl ester, benzoic acid,3-amino-4-methylamino-, ethyl ester, benzoic acid, 3-amino-4-methylamino-, ethyl ester |
Formula Weight | 194.23 |
Percent Purity | 97% |
Chemical Name or Material | ethyle3-amino-4-(methylamino)benzoate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ethyle3-amino-4-(methylamino)benzoate, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction