missing translation for 'onlineSavingsMsg'
Learn More

ethyle5-amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylate, 97%, Thermo Scientific™

Product Code. 10181272
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 138907-68-3
Molecular Formula C12H12FN3O2
Molecular Weight (g/mol) 249.25
MDL Number MFCD00173917
InChI Key RPPPCKSHIYWAPW-UHFFFAOYSA-N
Synonym ethyl 5-amino-1-4-fluorophenyl-1h-pyrazole-4-carboxylate, ethyl 5-amino-1-4-fluorophenyl pyrazole-4-carboxylate, ethyl 5-amino-1-4-fluoro-phenyl-1h-pyrazole-4-carboxylate, 5-amino-1-4-fluorophenyl-1h-pyrazole-4-carboxylic acid ethyl ester, 5-amino-4-ethoxycarbonyl-1-4-fluorophenyl-1h-pyrazole, 1-5-amino-4-ethoxycarbonyl-1h-pyrazol-1-yl-4-fluorobenzene, maybridge1_006170, ethyl5-amino-1-4-fluorophenyl-1h-pyrazole-4-carboxylate, ethylaminofluorophenylpyrazolecarboxylate, 5-amino-1-4-fluorophenyl-1h-pyrazole-4-carboxyli
PubChem CID 688689
IUPAC Name ethyl 5-amino-1-(4-fluorophenyl)pyrazole-4-carboxylate
SMILES CCOC(=O)C1=C(N)N(N=C1)C1=CC=C(F)C=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 249.24
Percent Purity 97%
Chemical Name or Material ethyle5-amino-1-(4-fluorophenyl)-1H-pyrazole-4-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'