missing translation for 'onlineSavingsMsg'
Learn More

ethyle6-iodo-4-oxo-4H-chromene-2-carboxylate, 97%, Thermo Scientific™

Product Code. 10522272
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 35204-44-5
Molecular Formula C12H9IO4
Molecular Weight (g/mol) 344.10
MDL Number MFCD00100378
InChI Key HUJFMOAXSPSGDU-UHFFFAOYSA-N
Synonym ethyl 6-iodo-4-oxo-4h-chromene-2-carboxylate, 4h-1-benzopyran-2-carboxylicacid, 6-iodo-4-oxo-, ethyl ester, maybridge1_002002, ethyl-6-iodo-4-oxo-4h-chromene-2-carboxylate, 4-oxo-6-iodo-4h-1-benzopyran-2-carboxylic acid ethyl ester
PubChem CID 2775235
IUPAC Name ethyl 6-iodo-4-oxo-4H-chromene-2-carboxylate
SMILES CCOC(=O)C1=CC(=O)C2=CC(I)=CC=C2O1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 344.1
Percent Purity 97%
Chemical Name or Material Ethyl 6-iodo-4-oxo-4H-chromene-2-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'