Learn More
Ethylene carbonate, 99+%, Thermo Scientific Chemicals
£16.80 - £144.00
Chemical Identifiers
CAS | 96-49-1 |
---|---|
Molecular Formula | C3H4O3 |
Molecular Weight (g/mol) | 88.06 |
MDL Number | MFCD00005382 |
InChI Key | KMTRUDSVKNLOMY-UHFFFAOYSA-N |
Synonym | ethylene carbonate, glycol carbonate, ethylene glycol carbonate, dioxolone-2, cyclic ethylene carbonate, ethylene carbonic acid, 1,3-dioxacyclopentan-2-one, 2-dioxolone, carbonic acid, cyclic ethylene ester, texacar ec |
PubChem CID | 7303 |
IUPAC Name | 1,3-dioxolan-2-one |
SMILES | C1COC(=O)O1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10724231
|
Thermo Scientific Acros
118412500 |
250 g | Glass bottle |
£16.80
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10539600
|
Thermo Scientific Acros
118410010 |
1 kg | Glass bottle |
£41.30
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10659853
|
Thermo Scientific Acros
118410051 |
5 kg | Plastic drum |
£144.00
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Alkylation reagentChemical Identifiers
96-49-1 | |
88.06 | |
KMTRUDSVKNLOMY-UHFFFAOYSA-N | |
7303 | |
C1COC(=O)O1 |
C3H4O3 | |
MFCD00005382 | |
ethylene carbonate, glycol carbonate, ethylene glycol carbonate, dioxolone-2, cyclic ethylene carbonate, ethylene carbonic acid, 1,3-dioxacyclopentan-2-one, 2-dioxolone, carbonic acid, cyclic ethylene ester, texacar ec | |
1,3-dioxolan-2-one |
Specifications
96-49-1 | |
White to Yellow | |
246.7°C | |
Authentic | |
C3H4O3 | |
19, 100 | |
1.321 | |
Solubility in water: 214g/L (20°C). Other solubilities: soluble in ether,n-butanol and carbon tetra-,chloride,miscible with alcohol,ethyl acetate,benzene and,chloroform | |
C1COC(=O)O1 | |
88.06 | |
1.92 mPa.s (40°C) | |
99+% | |
Ethylene carbonate |
34.0°C to 37.0°C | |
1.3210g/mL | |
150°C | |
99% min. (GC) | |
MFCD00005382 | |
01,372 | |
ethylene carbonate, glycol carbonate, ethylene glycol carbonate, dioxolone-2, cyclic ethylene carbonate, ethylene carbonic acid, 1,3-dioxacyclopentan-2-one, 2-dioxolone, carbonic acid, cyclic ethylene ester, texacar ec | |
KMTRUDSVKNLOMY-UHFFFAOYSA-N | |
1,3-dioxolan-2-one | |
7303 | |
88.06 | |
Crystalline Low Melting Solid |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 202-510-
- Ethylene carbonate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only