missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene chlorophosphite, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Ethylene chlorophosphite acts as a phosphitylating agent involved in the preparation of phosphocholine and spirophosphoranes. For example, it is used in the phosphorylation of p-tert-butyl(thia)calixarenes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethylene chlorophosphite acts as a phosphitylating agent involved in the preparation of phosphocholine and spirophosphoranes. For example, it is used in the phosphorylation of p-tert-butyl(thia)calixarenes.
Notes
Store in cool place. Moisture sensitive. Incompatible with water.
Ethylene chlorophosphite acts as a phosphitylating agent involved in the preparation of phosphocholine and spirophosphoranes. For example, it is used in the phosphorylation of p-tert-butyl(thia)calixarenes.
Notes
Store in cool place. Moisture sensitive. Incompatible with water.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 1.415 |
Boiling Point | 46°C to 47°C (15 mmHg) |
Flash Point | 86°C (186°F) |
Refractive Index | 1.491 |
Quantity | 25 g |
UN Number | UN3265 |
Beilstein | 506179 |
Sensitivity | Moisture sensitive |
Synonym | ethylene chlorophosphite, 1,3,2-dioxaphospholane, 2-chloro, ethylene cyclo-chlorophosphite, acmc-209pnx, 1,3,2-dioxaphospholane,2-chloro, 2-chloro-1,3,2-dioxaphospholane, ethylene phosphorochloridite 2-chloro-1,3,2-dioxap, inchi=1/c2h4clo2p/c3-6-4-1-2-5-6/h1-2h |
Formula Weight | 126.47 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'