missing translation for 'onlineSavingsMsg'
Learn More
Ethylene glycol, 99.5%, for analysis, Thermo Scientific Chemicals Product Code.: 10297410

Ethylene glycol, 99.5%, for analysis, Thermo Scientific Chemicals

Product Code. 10297410
2.5 L, Glass Bottle
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal Drum
This item is not returnable. View return policy

Product Code. 10297410

Brand: Thermo Scientific Acros 295530025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
MDL Number MFCD00002885
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescolShow MoreShow Less
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

Melting Point -13.0°C
Density 1.1130g/mL
Boiling Point 196.0°C to 198.0°C
Quantity 2.5 L
Flash Point 116°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Physical Form Viscous Liquid
Refractive Index 1.4300 to 1.4330
Assay Percent Range 99.5%
Specific Gravity 1.113
Linear Formula HOCH2CH2OH
Viscosity 21 mPa.s (20°C)
Beilstein 01, 465
Water 0.1% max. (K.F.)
Fieser 01,375; 05,296; 09,217; 15,156
Merck Index 15, 3852
Solubility Information Solubility in water: miscible. Other solubilities: miscible with acetic acid, acetone, glycerol,, pyridine, lower aliphatic alcohols,, aldehydes and ketones, slightly soluble in ether, practically insoluble in benzene and hydrocarbons
Formula Weight 62.06
Percent Purity 99.5%
Grade Analytical
Chemical Name or Material Ethylene glycol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1920718 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog Number295530250,295530010,295530025
Lot Number1988887 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number295530250,295530010,295530025
Lot Number1990406 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number295530250,295530010,295530025
Lot Number1902507 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number295530250,295530010,295530025
Lot Number1998916 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog Number295530250,295530010,295530025
compliance-icons compliance-icons
Product Identifier
  • Ethylene glycol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethylene glycol, 99.5%, for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.