missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol-d{6}, 98% (Isotopic)

Product Code. p-7051941
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11384328

Brand: Thermo Scientific Alfa Aesar 042288.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 15054-86-1 | C2H6O2 | 68.11 g/mol

It is used in synthetic materials and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15054-86-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 68.11
MDL Number MFCD00078666
InChI Key LYCAIKOWRPUZTN-UFSLNRCZSA-N
Synonym ethylene glycol-d6, 1,1,2,2-2h4 ethane-1,2-2h2 diol, 1,2-ethane-1,1,2,2-d4-diol-d2, 2 h? ethane-1,2-di 2 h ol, ethylen-d4 glycol-d2, ethylene glycol d6, ethylene glycol-d6, 98 atom % d, ethylene glycol-d6 >99.0 atom % d
PubChem CID 2733139
IUPAC Name (²H₄)ethane-1,2-di(²H)ol
SMILES [2H]OC([2H])([2H])C([2H])([2H])O[2H]

Specifications

Physical Form Liquid
Density 1.189 g/mL
Flash Point 111°C (232°F)
Odor Odorless
Quantity 1 g
Boiling Point 86°C to 87°C (8mmHg)
Solubility Information Soluble in water,methanol,DMSO
Formula Weight 68.02
Percent Purity 98%
Chemical Name or Material Ethylene glycol-d6, Isotopic
compliance-icons
Product Identifier
  • Ethylene glycol-d(6)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Ethylene glycol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.