missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene glycol diacetate, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
2500 g
Description
EGDA imparts excellent flow properties in baking lacquers and enamels and where thermoplastic acrylic resins are used. It is also a good solvent for cellulosic coatings and can be used in some ink systems such as screen inks. It has found use as a perfume fixative, and has reported applications in waterborne adhesives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | -41°C |
Density | 1.13 g/mL |
Boiling Point | 186°C to 187°C |
Flash Point | 82°C (181°F) |
Refractive Index | 1.4152 |
Quantity | 2500 g |
Beilstein | 1762308 |
Merck Index | 14,3799 |
Solubility Information | Soluble in water (16.4% by weight) |
Formula Weight | 146.14 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'