Learn More
Ethylene glycol diacetate, 97%, Thermo Scientific Chemicals
POA
Chemical Identifiers
CAS | 111-55-7 |
---|---|
Molecular Formula | C6H10O4 |
Molecular Weight (g/mol) | 146.142 |
MDL Number | MFCD00008718 |
InChI Key | JTXMVXSTHSMVQF-UHFFFAOYSA-N |
Synonym | ethylene glycol diacetate, 1,2-diacetoxyethane, glycol diacetate, ethylene diacetate, ethanediol diacetate, 1,2-ethanediol, diacetate, 1,2-ethanediol diacetate, ethylene glycol, diacetate, ethylene diethanoate, aptex donor h-plus |
PubChem CID | 8121 |
IUPAC Name | 2-acetyloxyethyl acetate |
SMILES | CC(=O)OCCOC(=O)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15421737
|
Thermo Scientific Alfa Aesar
041910.0E |
2500 g | |||||||
Description
EGDA imparts excellent flow properties in baking lacquers and enamels and where thermoplastic acrylic resins are used. It is also a good solvent for cellulosic coatings and can be used in some ink systems such as screen inks. It has found use as a perfume fixative, and has reported applications in waterborne adhesives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
111-55-7 | |
146.142 | |
JTXMVXSTHSMVQF-UHFFFAOYSA-N | |
8121 | |
CC(=O)OCCOC(=O)C |
C6H10O4 | |
MFCD00008718 | |
ethylene glycol diacetate, 1,2-diacetoxyethane, glycol diacetate, ethylene diacetate, ethanediol diacetate, 1,2-ethanediol, diacetate, 1,2-ethanediol diacetate, ethylene glycol, diacetate, ethylene diethanoate, aptex donor h-plus | |
2-acetyloxyethyl acetate |
Specifications
111-55-7 | |
1.13 g/mL | |
82°C (181°F) | |
1.4152 | |
1762308 | |
ethylene glycol diacetate, 1,2-diacetoxyethane, glycol diacetate, ethylene diacetate, ethanediol diacetate, 1,2-ethanediol, diacetate, 1,2-ethanediol diacetate, ethylene glycol, diacetate, ethylene diethanoate, aptex donor h-plus | |
JTXMVXSTHSMVQF-UHFFFAOYSA-N | |
2-acetyloxyethyl acetate | |
8121 | |
97% | |
Ethylene glycol diacetate |
-41°C | |
186°C to 187°C | |
C6H10O4 | |
MFCD00008718 | |
14,3799 | |
Soluble in water (16.4% by weight) | |
CC(=O)OCCOC(=O)C | |
146.142 | |
146.14 | |
Liquid |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
missing translation for 'einecsNumber' : 203-881-1
missing translation for 'rtecsNumber' : KW4025000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only