Learn More
Ethylene glycol dimethyl ether, 99.5%, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 442980010
Code : 29
Additional Details : CAS Number : 110-71-4 Weight : 0.86700kg Transport : UN number : 2252 Chem class : 3 Pack group : II
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethylene glycol dimethyl ether | |
110-71-4 | |
84.0°C to 86.0°C | |
−6°C | |
0.867 | |
1.1 mPa.s (20°C) | |
monoglyme, ethylene glycol dimethyl ether, glyme, dimethyl cellosolve, egdme, ethane, 1,2-dimethoxy, dimethoxyethane, 2,5-dioxahexane, glycol dimethyl ether, ethylene dimethyl ether | |
90.12 | |
Extra Dry over Molecular Sieve |
-69.0°C | |
0.8670g/mL | |
1 L | |
1.3785 to 1.3805 | |
CH3OCH2CH2OCH3 | |
0.005% max. | |
Solubility in water: miscible. Other solubilities: solulbe in hydrocarbon solvents, soluble in ethanol, ethyl ether, acetone, carbon, tetrachloride, benzene, dichloroethane | |
99.5% |
Safety and Handling
- Ethylene glycol dimethyl ether
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Flammable liquid Category 2
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
Hazard Statement
- H360FD-May damage fertility. May damage the unborn child.
- H315-Causes skin irritation.
- H332-Harmful if inhaled.
- H225-Highly flammable liquid and vapour.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only