missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol dimethyl ether, 99%, Extra Dry, AcroSeal(R), Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 610351000

38.23 GBP valid until 2024-12-31
Use promo code "21665" to get your promotional price.



 View more versions of this product

Product Code. 10336744

  • £44.02 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Ethylene glycol dimethyl ether
128-37-0
84.0°C to 86.0°C
-6°C (-42.8°F)
AcroSeal™ Glass Bottle
99%
Vapor Pressure: 64hPa at 20°C
50ppm max.
15, 3245
90.12
Extra Dry
-69°C
0.8670g/mL
100 mL
Pass Test
1.3785 to 1.3805
CH3OCH2CH2OCH3
01, 467
01,267; 02,143; 16,229
monoglyme, ethylene glycol dimethyl ether, glyme, dimethyl cellosolve, egdme, ethane, 1,2-dimethoxy, dimethoxyethane, 2,5-dioxahexane, glycol dimethyl ether, ethylene dimethyl ether
≥99%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ethylene glycol dimethyl ether

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
  • H315-Causes skin irritation.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

RUO – Research Use Only