missing translation for 'onlineSavingsMsg'
Learn More

Ethylene Glycol, Fisher BioReagents™

Product Code. 10532595
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle

Ethylene Glycol

Due to its property as an antifreeze, Ethylene Glycol may be used for the storage of enzymes at low temperatures.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

pH 5.5 to 7.5
Melting Point -13°C
Color Colorless
Decomposition Information >500°C
Density 1.113g/cm³
Boiling Point 196°C
Quantity 4 L
Identification Pass Test
Packaging Glass Bottle
Physical Form Viscous Liquid
Assay Percent Range ≥99 %
Titratable Acid 0.002mEq/g max.
Linear Formula HOCH2CH2OH
Vapor Pressure 0.12mmHg at 20°C
Viscosity 21 cP at 20°C
Water 0.5% max.
Merck Index 15, 3852
Formula Weight 62.06
Percent Purity ≥99%
Chemical Name or Material Ethylene Glycol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethylene glycol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.