missing translation for 'onlineSavingsMsg'
Learn More

Ethylene glycol, Reagent Grade, ≥99%, Honeywell™

Product Code. 15600970 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
6 x 500 mL
20 L
30 kg
4 x 2.5 L
225 kg
Packaging:
Metal Drum
Poly Bottle
Stainless Steel Can
Unit Size:
1 L
2.5 L
20 L
225kg
30kg
5 L
500mL
Pack of 4
Pack of 6
This item is not returnable. View return policy

Product Code. 15600970

Brand: Honeywell Research Chemicals 102466500ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-21-1
Molecular Formula C2H6O2
Molecular Weight (g/mol) 62.068
MDL Number MFCD00002885
InChI Key LYCAIKOWRPUZTN-UHFFFAOYSA-N
Synonym ethylene glycol, 1,2-ethanediol, glycol, monoethylene glycol, 1,2-dihydroxyethane, 2-hydroxyethanol, glycol alcohol, ethylene alcohol, fridex, tescol
PubChem CID 174
ChEBI CHEBI:30742
IUPAC Name ethane-1,2-diol
SMILES C(CO)O

Specifications

Melting Point -13°C (literature)
CAS Max % 100.0000%
Density 1.113 g/mL at 25 °C(lit.)
Boiling Point 197°C
Quantity 500 mL
Flash Point 111°C
Packaging Poly Bottle
Linear Formula HOCH2CH2OH
Beilstein 505945
Formula Weight 62.07
Percent Purity >99%
Grade Reagent
Chemical Name or Material Ethylene glycol, Reagent Grade, ≥99%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.