missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene glycol, Reagent Grade, ≥99%, Honeywell™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
6 x 500 mL
20 L
30 kg
4 x 2.5 L
225 kg
Packaging:
Metal Drum
Poly Bottle
Stainless Steel Can
Description
- ≥95% for general laboratory use.

Specifications
Specifications
Melting Point | -13°C (literature) |
CAS Max % | 100.0000% |
Density | 1.113 g/mL at 25 °C(lit.) |
Boiling Point | 197°C |
Quantity | 5 L |
Explosion Limit | 0.153 |
Flash Point | 111°C |
Ignition Point | 752°F |
Packaging | Poly Bottle |
Linear Formula | HOCH2CH2OH |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Safety and Handling


Product Identifier
- Ethylene glycol
- Warning
- Acute toxicity Category 4
- Specific target organ toxicity after repeated exposure Category 2
- H302-Harmful if swallowed.
- H373-May cause damage to organs.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : Ethylene glycol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ethylene glycol, Reagent Grade, ≥99%, Honeywell™ >
Spot an opportunity for improvement?Share a Content Correction