missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediamine, 99%, Thermo Scientific Chemicals

Product Code. 10244960
Click to view available options
Quantity:
5 mL
1 L
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-15-3
Molecular Formula C2H8N2
Molecular Weight (g/mol) 60.10
MDL Number MFCD00008204
InChI Key PIICEJLVQHRZGT-UHFFFAOYSA-N
Synonym ethylenediamine, 1,2-ethanediamine, 1,2-diaminoethane, ethylene diamine, ethylenediamine, dimethylenediamine, edamine, 1,2-ethylenediamine, aethaldiamin, aethylenediamin
PubChem CID 3301
ChEBI CHEBI:30347
IUPAC Name ethane-1,2-diamine
SMILES NCCN

Specifications

Color Colorless to Yellow
Density 0.8980g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula H2NCH2CH2NH2
Refractive Index 1.4555 to 1.4575 (20°C, 589nm)
Quantity 5 mL
Beilstein 04, 230
Fieser 01,372; 04,231; 13,157
Merck Index 15, 3849
Formula Weight 60.1
Percent Purity ≥98.5%
Grade Extra Pure
Physical Form Liquid, Fuming In Air
Chemical Name or Material Ethylenediamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'