Learn More
Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, 99%, pure
£29.85 - £283.41
Chemical Identifiers
CAS | 60-00-4 |
---|---|
Molecular Formula | C10H16N2O8 |
Molecular Weight (g/mol) | 292.24 |
MDL Number | MFCD00003541 |
InChI Key | KCXVZYZYPLLWCC-UHFFFAOYSA-N |
Synonym | edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol |
PubChem CID | 6049 |
ChEBI | CHEBI:42191 |
IUPAC Name | 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid |
SMILES | OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10264960
|
Thermo Scientific Chemicals
118432500 |
250 g | Plastic bottle |
£29.85
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10704611
|
Thermo Scientific Chemicals
118430010 |
1 kg | Plastic bottle |
£80.30
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11955331
|
Thermo Scientific Chemicals
118430050 |
5 kg | Plastic drum |
£283.41
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
60-00-4 | |
292.24 | |
KCXVZYZYPLLWCC-UHFFFAOYSA-N | |
6049 | |
2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid |
C10H16N2O8 | |
MFCD00003541 | |
edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol | |
CHEBI:42191 | |
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O |
Specifications
60-00-4 | |
White | |
>100°C | |
98.5% min. (Complexometry) | |
(HO2CCH2)2NCH2CH2N(CH2CO2H)2 | |
01,373 | |
edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol | |
KCXVZYZYPLLWCC-UHFFFAOYSA-N | |
2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid | |
6049 | |
292.23 | |
Pure | |
Ethylenediaminetetraacetic acid |
220.0°C | |
∼2 to 3 (sat. soln. aq. soln.) | |
Authentic | |
C10H16N2O8 | |
MFCD00003541 | |
15,3565 | |
Solubility in water: 0.5g/L (20°C) - 2.2g/L (80°C). Other solubilities: insoluble in common organic solvents, insoluble in dilute ammonium hydroxide: 0.005% max | |
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O | |
292.24 | |
CHEBI:42191 | |
99% | |
Fine Crystalline Powder |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES: Rinse cau
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 200-449-4
- Ethylenediaminetetraacetic acid
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Acute toxicity Category 4
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H373-May cause damage to organs.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.