missing translation for 'onlineSavingsMsg'
Learn More
Ethylenediaminetetraacetic acid, ACS, 99.4+%, Thermo Scientific Chemicals Product Code.: 11386378

Ethylenediaminetetraacetic acid, ACS, 99.4+%, Thermo Scientific Chemicals

Product Code. 11386378
500 g
Click to view available options
Quantity:
100 g
500 g
2.5 kg
This item is not returnable. View return policy

Product Code. 11386378

Brand: Thermo Scientific Alfa Aesar 011931.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chelating agent

Chelating agentEthylenediaminetetraacetic acid is used as a complexing agent for many cations. It prevents metal ion impurities and is used to remove excess iron from the body. It acts as a sequestering agent in cosmetic products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60-00-4
Molecular Formula C10H16N2O8
Molecular Weight (g/mol) 292.24
MDL Number MFCD00003541
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
PubChem CID 6049
ChEBI CHEBI:42191
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

Specifications

Melting Point ∼245°C (decomposition)
Color White
Linear Formula (HO2CCH2)2NCH2CH2N(CH2CO2H)2
Quantity 500 g
Beilstein 1716295
Merck Index 14,3517
Solubility Information Soluble in sodium hydroxide,sodium carbonate,ammonia solution,acid and common organic solvents. Slightly soluble in water.
Formula Weight 292.24
Percent Purity ≥99.4%
Grade ACS
Physical Form Powder
Chemical Name or Material Ethylenediaminetetraacetic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethylenediaminetetraacetic acid, ACS
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Ethylenediaminetetraacetic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethylenediaminetetraacetic acid, ACS, 99.4+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.