missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediaminetetraacetic acid copper(II) disodium salt solution, Honeywell™
Click to view available options
Quantity:
500 mL
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
CAS | 39208-15-6 |
Quantity | 500 mL |
Molecular Formula | C10H20CuN2Na2O12 |
MDL Number | MFCD00148920 |
UN Number | UN3082 |
Synonym | copper ii disodium ethylenediaminetetraacetate tetrahydrate, ethylenediaminetetraacetic acidcopper ii disodium salt tetrahydrate, acmc-209j3w, edta-cu-15, ksc911m0f, copper 2+ disodium ion 4-edta tetrahydrate, copper ii disodium 2,2',2,2'-ethane-1,2-diylbis azanetriyl tetraacetate tetrahydrate |
InChI Key | CHPMNDHAIUIBSK-UHFFFAOYSA-J |
SMILES | C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].O.O.O.O.[Na+].[Na+].[Cu+2] |
IUPAC Name | copper;disodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;tetrahydrate |
Molecular Weight (g/mol) | 469.798 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'