missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediaminetetraacetic Acid Disodium Magnesium Salt Tetrahydrate, Honeywell Fluka™
Click to view available options
Quantity:
100 g
Specifications
CAS | 29932-54-5 |
Quantity | 100 g |
Molecular Formula | C10H20MgN2Na2O12 |
Linear Formula | (NaOOCCH2)2NCH2CH2N(CH2COO)2Mg · 4H2O |
UN Number | NONH for all modes of transport |
Synonym | ethylenediaminetetraacetic acid disodium magnesium salt tetrahydrate |
InChI Key | NTLBRFLUZXNZAU-UHFFFAOYSA-J |
SMILES | C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].O.O.O.O.[Na+].[Na+].[Mg+2] |
IUPAC Name | magnesium;disodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;tetrahydrate |
Molecular Weight (g/mol) | 430.557 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'