missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediaminetetraacetic Acid Disodium Salt Dihydrate, ≥99%, Molecular Biology Reagent Grade, MP Biomedicals™
Click to view available options
Quantity:
500 g
Specifications
Color | White |
Assay Percent Range | ≥99% |
Quantity | 500 g |
IUPAC Name | disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate |
Formula Weight | 372.2 |
Grade | Molecular Biology Reagent |
Physical Form | Crystalline Powder |
Chemical Name or Material | Ethylenediaminetetraacetic Acid Disodium Salt Dihydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'