missing translation for 'onlineSavingsMsg'
Learn More
Ethylenediaminetetraacetic Acid Disodium Salt Solution 0.01M (0.02N), NIST Standard Concentrate, for volumetric analysis, Fisher Chemical Product Code.: 10497060

Ethylenediaminetetraacetic Acid Disodium Salt Solution 0.01M (0.02N), NIST Standard Concentrate, for volumetric analysis, Fisher Chemical

Product Code. 10497060
6 Ampules, Ampoule
Click to view available options
Quantity:
6 Ampules
Packaging:
Ampoule
This item is not returnable. View return policy

Product Code. 10497060

Brand: Fisher Chemical J/3720C/90

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Specifications

CAS 139-33-3
Color Colorless
Packaging Ampoule
Molecular Formula C10H14N2Na2O8
MDL Number MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
Quantity 6 Ampules
Synonym ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid
InChI Key ZGTMUACCHSMWAC-UHFFFAOYSA-L
SMILES [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
IUPAC Name disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate
Molecular Weight (g/mol) 336.21
PubChem CID 57339238
ChEBI CHEBI:64734
Physical Form Liquid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2525939 Certificate TypeCertificate of Analysis Date10/02/2025 Catalog NumberJ/3720C/90,J/3720C/05
Product Identifier
  • EDTA (Diaminoethanetetra-acetic acid, disodium salt solution 0.1 M) (0.2N)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethylenediaminetetraacetic Acid Disodium Salt Solution 0.01M (0.02N), NIST Standard Concentrate, for volumetric analysis, Fisher Chemical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.