missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediaminetetraacetic Acid Disodium Salt Solution 0.1M (0.2N), NIST Standard Solution ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP , Fisher Chemical
Brand: Fisher Chemical J/3700/15
50.60 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
1L, HDPE plastic bottle
Ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP
Specifications
139-33-3 | |
HDPE plastic bottle | |
MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037 | |
ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid | |
[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O | |
336.21 | |
CHEBI:64734 |
Colorless | |
C10H14N2Na2O8 | |
1 L | |
ZGTMUACCHSMWAC-UHFFFAOYSA-L | |
disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate | |
57339238 | |
Liquid |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Safety and Handling
Product Identifier
- EDTA (Diaminoethanetetra-acetic acid, disodium salt solution 0.1 M) (0.2N)
Fisherbrand Focus on pH and electrochemistry
File size: 9.1 MB
Fisherbrand Focus on pH and electrochemistry
File size: 2.5 MB
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2525939 | Certificate of Analysis | 06/02/2025 | J/3700C/05,J/3700C/90,J/3700/15,J/3700/24,J/3700/17 |
Special Offers
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ethylenediaminetetraacetic Acid Disodium Salt Solution 0.1M (0.2N), NIST Standard Solution ready to use, for volumetric analysis, meets analytical specification of Ph.Eur., BP , Fisher Chemical > 1L, HDPE plastic bottle
Spot an opportunity for improvement?Share a Content Correction