missing translation for 'onlineSavingsMsg'
Learn More

EDTA disodium salt solution, Volumetric, Reag. Ph. Eur., 0.1 M EDTA-Na2, for complexometry, Honeywell Fluka™

Volumetric, Reag. Ph. Eur., 0.1 M EDTA-Na2, for complexometry

£30.46 - £386.78

Specifications

CAS 139-33-3
Molecular Formula C10H14N2Na2O8
MDL Number MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
Synonym ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid
InChI Key ZGTMUACCHSMWAC-UHFFFAOYSA-L
View More Specs

Products 9
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15614280
View Documents Promotion Details
Honeywell-Fluka
34550-500ML
500 mL
£46.34
500mL
Please to purchase this item. Need a web account? Register with us today!
15694270
View Documents
Honeywell-Fluka
34550-1L
1 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15604280
View Documents
Honeywell-Fluka
34550-2.5L
2.5 L
£122.84
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15654280
View Documents
Honeywell-Fluka
34550-6X500ML
6 x 500 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15635735
View Documents
Honeywell-Fluka
34550-5L-VP
5 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15624280
View Documents
Honeywell-Fluka
34550-5L
5 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15644280
View Documents
Honeywell-Fluka
34550-6X1L
6 x 1 L
£293.82
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
15625735
View Documents
Honeywell-Fluka
34550-10L-VP
10 L
£386.78
10 L
Please to purchase this item. Need a web account? Register with us today!
15674270
View Documents
Honeywell-Fluka
34550-10L
10 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

Specifications

Specifications

139-33-3
MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037
ZGTMUACCHSMWAC-UHFFFAOYSA-L
336.21
CHEBI:64734
C10H14N2Na2O8
ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid
[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
57339238
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
EDTA disodium salt solution, Volumetric, Reag. Ph. Eur., 0.1 M EDTA-Na2, for complexometry, Honeywell Fluka™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.