missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediaminetetraacetic Acid Disodium Salt Solution, 0.2M, Honeywell Fluka™
217.05 GBP valid until 2024-12-31
Use promo code "21986" to get your promotional price.
Use promo code "21986" to get your promotional price.
Click to view available options
Quantity:
1 L
5 L
10 L
6 x 1 L
Packaging:
Cubitainer
Poly Bottle
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Linear Formula | [-CH2N(CH2CO2Na)CH2CO2H]2 |
IUPAC Name | disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate |
Formula Weight | 336.21g/mol |
Packaging | Cubitainer |
Quantity | 5 L |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'