missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediaminetetraacetic Acid Disodium Salt Solution, 0.2M, Honeywell Fluka™
Click to view available options
Quantity:
1 L
5 L
10 L
6 x 1 L
Packaging:
Cubitainer
Poly Bottle
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Chemical Name or Material | Ethylenediaminetetraacetic acid disodium salt solution |
Molecular Formula | C10H14N2Na2O8 |
Linear Formula | [-CH2N(CH2CO2Na)CH2CO2H]2 |
MDL Number | MFCD00070672,MFCD00003541,MFCD00070672,MFCD00150037 |
UN Number | NONH for all modes of transport |
Beilstein | 3822669 |
Synonym | ethylenediaminetetraacetic acid disodium salt, edta na2, disodium 2-2-bis carboxymethyl amino ethyl-carboxymethyl amino acetic acid |
InChI Key | ZGTMUACCHSMWAC-UHFFFAOYSA-L |
SMILES | [Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O |
IUPAC Name | disodium;2-[2-[bis(carboxylatomethyl)azaniumyl]ethyl-(carboxylatomethyl)azaniumyl]acetate |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'