Learn More
Honeywell Fluka™ EDTA disodium salt solution with zinc complex added (solution A), Volumetric, for water hardness determination, Honeywell Fluka™
Volumetric, for water hardness determination (1 ml = 5.6 German degree of hardness in 100 ml of water)
Brand: Honeywell Fluka™ 34547-6X1L
Code : NEW
Additional Details : Weight : 6.00000kg
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Poly Bottle | |
6 X 1 L | |
Room Temperature | |
EDTA disodium salt solution with zinc complex added (solution A) |