missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EDTA disodium salt solution with zinc complex added (solution B), 1 ml solution = 1 German degree of hardness in 100 ml of water, for complexometry, Honeywell Fluka™
Click to view available options
Quantity:
1 L
5 L
6 x 500 mL
10 L
6 x 1 L
Packaging:
Cubitainer
Poly Bottle
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Chemical Name or Material | Ethylenediaminetetraacetic acid disodium salt solution with zinc complex added (solution B) |
CAS | 7646-85-7, 60-00-4, 7732-18-5 |
CAS Max % | 0.7, 0.02 |
Molecular Formula | Cl2Zn |
UN Number | NONH for all modes of transport |
InChI Key | JIAARYAFYJHUJI-UHFFFAOYSA-L |
SMILES | [Cl-].[Cl-].[Zn++] |
IUPAC Name | zinc(2+) dichloride |
Molecular Weight (g/mol) | 136.28 |
Packaging | Poly Bottle |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'