Learn More
Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate, 98%, pure
Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate, 98%, C10H12FeN2NaO8.3H2O, CAS Number-18154-32-, sodium ironedetate, edta ferric sodium salt, calmosine, sodium feredetate, edta iron iii sodium salt, sequestrene nafe, ferric sodium edta, ferisan, sodium iron edta, sytron, 1kg
£109.00 - £992.64
Chemical Identifiers
CAS | 18154-32-0 |
---|---|
Molecular Formula | C10H12FeN2NaO8·3H2O |
Molecular Weight (g/mol) | 421.1 |
MDL Number | MFCD00798117 |
InChI Key | MKWYFZFMAMBPQK-UHFFFAOYSA-J |
Synonym | sodium feredetate, edta ferric sodium salt, calmosine, sodium ironedetate, edta iron iii sodium salt, ferisan, sytron, sequestrene nafe, sodium iron edta, ferric sodium edta |
PubChem CID | 27461 |
ChEBI | CHEBI:78292 |
IUPAC Name | sodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;iron(3+) |
SMILES | C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].[Na+].[Fe+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10175750
|
Thermo Scientific Chemicals
304682500 |
250 g |
£109.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10287320
|
Thermo Scientific Chemicals
304680010 |
1 kg |
£331.00
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10677554
|
Thermo Scientific Chemicals
304680051 |
5 kg | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagentChemical Identifiers
18154-32-0 | |
421.1 | |
MKWYFZFMAMBPQK-UHFFFAOYSA-J | |
27461 | |
sodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;iron(3+) |
C10H12FeN2NaO8·3H2O | |
MFCD00798117 | |
sodium feredetate, edta ferric sodium salt, calmosine, sodium ironedetate, edta iron iii sodium salt, ferisan, sytron, sequestrene nafe, sodium iron edta, ferric sodium edta | |
CHEBI:78292 | |
C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].[Na+].[Fe+3] |
Specifications
18154-32-0 | |
Authentic | |
Plastic bottle | |
MFCD00798117 | |
sodium feredetate, edta ferric sodium salt, calmosine, sodium ironedetate, edta iron iii sodium salt, ferisan, sytron, sequestrene nafe, sodium iron edta, ferric sodium edta | |
MKWYFZFMAMBPQK-UHFFFAOYSA-J | |
sodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;iron(3+) | |
27461 | |
421.1 | |
Pure | |
Ethylenediaminetetraacetic acid, ferric sodium salt trihydrate, 98% |
Brown-Yellow | |
98% | |
C10H12FeN2NaO8·3H2O | |
15,4058 | |
Solubility in water: soluble. | |
C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].[Na+].[Fe+3] | |
421.1 | |
CHEBI:78292 | |
98% | |
Powder |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RUO – Research Use Only