Learn More
Thermo Scientific Chemicals Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate, 99%
£21.91 - £100.20
Chemical Identifiers
CAS | 10378-23-1 |
---|---|
Molecular Formula | C10H12N2Na4O8·2H2O |
Molecular Weight (g/mol) | 416.21 |
InChI Key | DGWOWDJDIBPYOR-UHFFFAOYSA-N |
Synonym | ethylenediaminetetraacetic acid tetrasodium salt dihydrate, tetrasodium ethylenediaminetetraacetate dihydrate, edta-na4, edta tetrasodium salt, edta tetrasodium salt hydrate, ethylenediaminetetraacetic acid tetrasodium salt hydrate, 64-02-8 anhydrous, ksc909a7h, edta tetrasodium salt dihydrate, tetrapotassium ion 4-edta dihydrate |
PubChem CID | 131843362 |
IUPAC Name | 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;sodium;dihydrate |
SMILES | C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.O.O.[Na].[Na].[Na].[Na] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12675017
|
Thermo Scientific Chemicals
446081000 |
100 g | Plastic bottle |
£21.91
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12685017
|
Thermo Scientific Chemicals
446085000 |
500 g | Plastic bottle |
£64.02
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12665017
|
Thermo Scientific Chemicals
446080010 |
1 kg | Plastic bottle |
£100.20
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagentChemical Identifiers
10378-23-1 | |
416.21 | |
ethylenediaminetetraacetic acid tetrasodium salt dihydrate, tetrasodium ethylenediaminetetraacetate dihydrate, edta-na4, edta tetrasodium salt, edta tetrasodium salt hydrate, ethylenediaminetetraacetic acid tetrasodium salt hydrate, 64-02-8 anhydrous, ksc909a7h, edta tetrasodium salt dihydrate, tetrapotassium ion 4-edta dihydrate | |
2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;sodium;dihydrate |
C10H12N2Na4O8·2H2O | |
DGWOWDJDIBPYOR-UHFFFAOYSA-N | |
131843362 | |
C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.O.O.[Na].[Na].[Na].[Na] |
Specifications
10378-23-1 | |
99.0 to 102% (on dry substance) (Complexometry) | |
ethylenediaminetetraacetic acid tetrasodium salt dihydrate, tetrasodium ethylenediaminetetraacetate dihydrate, edta-na4, edta tetrasodium salt, edta tetrasodium salt hydrate, ethylenediaminetetraacetic acid tetrasodium salt hydrate, 64-02-8 anhydrous, ksc909a7h, edta tetrasodium salt dihydrate, tetrapotassium ion 4-edta dihydrate | |
DGWOWDJDIBPYOR-UHFFFAOYSA-N | |
2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid;sodium;dihydrate | |
131843362 | |
99% |
300.0°C | |
C10H12N2Na4O8·2H2O | |
(16% in water) Clear solution | |
C(CN(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O.O.O.[Na].[Na].[Na].[Na] | |
416.21 | |
416.21 | |
Ethylenediaminetetraacetic acid, tetrasodium salt dihydrate |
Safety and Handling
GHS H Statement
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Danger
- Ethylenediaminetetraacetic acid tetrasodium salt dihydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only