missing translation for 'onlineSavingsMsg'
Learn More

Ethylenediaminetetraacetic Acid (White Crystalline Powd.), Fisher BioReagents™

Product Code. 10011123
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
Unit Size:
500g
This item is not returnable. View return policy

Product Code. 10011123

Brand: Fisher BioReagents BP118500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
EDTA is a chelating agent for metal ions and is used extensively in molecular biology to minimize metal ion impurities in reaction buffers.
TRUSTED_SUSTAINABILITY

Specifications

CAS 60-00-4
Melting Point 220°C
Color White
pH 2.5
Ignition Residue 0.2% max.
Assay Percent Range ≥99.5 %
Packaging Glass Bottle
Molecular Formula C10H16N2O8
Linear Formula (HOOCCH2)2NCH2CH2N(CH2COOH)2
MDL Number MFCD00003541
Quantity 500 g
Merck Index 15, 3565
Synonym edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol
InChI Key KCXVZYZYPLLWCC-UHFFFAOYSA-N
SMILES OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
IUPAC Name 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid
Molecular Weight (g/mol) 292.24
PubChem CID 6049
ChEBI CHEBI:42191
Formula Weight 292.23
Vapor Pressure 0.013 hPa at 20°C
Percent Purity ≥99.5%
Physical Form Powder or Solid
Chemical Name or Material Ethylenediamine Tetraacetic Acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethylenediaminetetraacetic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Acute toxicity Category 4
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.