missing translation for 'onlineSavingsMsg'
Learn More

Ethylidene diacetate, 99%, Thermo Scientific Chemicals

Product Code. 11914921
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 542-10-9
Molecular Formula C6H10O4
Molecular Weight (g/mol) 146.14
InChI Key ACKALUBLCWJVNB-UHFFFAOYSA-N
Synonym ethylidene diacetate, 1,1-diacetoxyethane, 1,1-ethanediol diacetate, 1,1-ethanediol, diacetate, ethylidene acetate, ethane-1,1-diyl diacetate, polyoxymethylenes, delrin, unii-kl1s8v6w25, 1-acetoxyethyl acetate
PubChem CID 222536
IUPAC Name 1-acetyloxyethyl acetate
SMILES CC(OC(=O)C)OC(=O)C

Specifications

Color Colorless to Yellow
Boiling Point 69.0°C (17.0 mmHg)
Flash Point 68°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula CH3CH(OCOCH3)2
Refractive Index 1.3995 to 1.4015
Quantity 500 g
Merck Index 15,3866
Formula Weight 146.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Ethylidene diacetate
Show More Show Less
Product Identifier
  • Ethylidene diacetate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'