missing translation for 'onlineSavingsMsg'
Learn More
Ethylmagnesium bromide, 0.9M solution in THF, AcroSeal™, Thermo Scientific Chemicals Product Code.: 10444833

Ethylmagnesium bromide, 0.9M solution in THF, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10444833
800 mL, AcroSeal™ Glass Bottle
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Product Code. 10444833

Brand: Thermo Scientific Acros 210478000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 925-90-6, 109-99-9
Molecular Formula C2H5BrMg
Molecular Weight (g/mol) 133.27
MDL Number MFCD00000043
InChI Key TWTWFMUQSOFTRN-UHFFFAOYSA-M
Synonym ethylmagnesium bromide, ethyl magnesium bromide, magnesium, bromoethyl, ethylmagnesiumbromide, bromoethylmagnesium, ethylmagnesium bromide solution, 1.0 m in thf, ethylmagnesium bromide solution, 3.0 m in diethyl ether, etmgbr, ethylmagnesiumbromid, ethylmagnesium bromid, grignard reagent
PubChem CID 101914
SMILES CC[Mg]Br

Specifications

Color Black-Brown to Brown
Linear Formula C2H5MgBr
Concentration 0.85 to 1.15M
Fieser 02,205; 08,510; 11,574
Density 1.0100g/mL
Solubility Information Solubility in water: vigorous reaction.
IUPAC Name bromo(ethyl)magnesium
Formula Weight 133.27
Packaging AcroSeal™ Glass Bottle
Specific Gravity 1.01
Quantity 800 mL
Flash Point −17°C
Physical Form Solution
Chemical Name or Material Ethylmagnesium bromide, 1M Solution in THF
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0443558 Certificate TypeCertificate of Analysis Date13/03/2024 Catalog Number210478000,210471000
Lot NumberA0459580 Certificate TypeCertificate of Analysis Date11/03/2024 Catalog Number210478000,210471000
Lot NumberA0395976 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number210478000,210471000
Lot NumberA0414142 Certificate TypeCertificate of Analysis Date20/05/2021 Catalog Number210478000,210471000
Lot NumberA0411228 Certificate TypeCertificate of Analysis Date10/02/2021 Catalog Number210478000,210471000
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethyl magnesiumbromide, 1M solution in THF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethylmagnesium bromide, 0.9M solution in THF, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.