Learn More
Ethylmagnesium bromide, 3M in ether, Thermo Scientific Chemicals
Grignard reagent
£69.60 - £108.00
Chemical Identifiers
CAS | 925-90-6 |
---|---|
Molecular Formula | C2H5BrMg |
Molecular Weight (g/mol) | 133.27 |
MDL Number | MFCD00000043 |
InChI Key | TWTWFMUQSOFTRN-UHFFFAOYSA-M |
Synonym | ethylmagnesium bromide, ethyl magnesium bromide, magnesium, bromoethyl, ethylmagnesiumbromide, bromoethylmagnesium, ethylmagnesium bromide solution, 1.0 m in thf, ethylmagnesium bromide solution, 3.0 m in diethyl ether, etmgbr, ethylmagnesiumbromid, ethylmagnesium bromid, grignard reagent |
PubChem CID | 101914 |
IUPAC Name | magnesium;ethane;bromide |
SMILES | CC[Mg]Br |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11460950
|
Thermo Scientific Alfa Aesar
087291.JT |
0.5 mol |
£69.60
0.50mol |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11450950
|
Thermo Scientific Alfa Aesar
087291.JU |
1 mol |
£108.00
1mol |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ethylmagnesium bromide is used as grignard reagent. It is widely used in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
925-90-6 | |
133.27 | |
TWTWFMUQSOFTRN-UHFFFAOYSA-M | |
101914 | |
CC[Mg]Br |
C2H5BrMg | |
MFCD00000043 | |
ethylmagnesium bromide, ethyl magnesium bromide, magnesium, bromoethyl, ethylmagnesiumbromide, bromoethylmagnesium, ethylmagnesium bromide solution, 1.0 m in thf, ethylmagnesium bromide solution, 3.0 m in diethyl ether, etmgbr, ethylmagnesiumbromid, ethylmagnesium bromid, grignard reagent | |
magnesium;ethane;bromide |
Specifications
925-90-6 | |
Brown to Yellow | |
34.6°C | |
C2H5BrMg | |
MFCD00000043 | |
Air and moisture sensitive; May form a precipitate | |
Reacts with water. | |
CC[Mg]Br | |
133.27 | |
133.28 | |
Liquid |
-116.3°C | |
1.020 g/mL | |
Ether-like | |
C2H5MgBr | |
UN3399 | |
ethylmagnesium bromide, ethyl magnesium bromide, magnesium, bromoethyl, ethylmagnesiumbromide, bromoethylmagnesium, ethylmagnesium bromide solution, 1.0 m in thf, ethylmagnesium bromide solution, 3.0 m in diethyl ether, etmgbr, ethylmagnesiumbromid, ethylmagnesium bromid, grignard reagent | |
TWTWFMUQSOFTRN-UHFFFAOYSA-M | |
magnesium;ethane;bromide | |
101914 | |
3M in ether | |
Ethylmagnesium bromide |
Safety and Handling
GHS H Statement
H224-H260-H314-H336
Extremely flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
May cause drowsiness or dizziness.
P210-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H224-H260-H302-H314-H336
missing translation for 'dotInformation' : Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
missing translation for 'einecsNumber' : 213-127-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only