missing translation for 'onlineSavingsMsg'
Learn More

Ethyltriphenylphosphonium bromide, 98%, Thermo Scientific Chemicals

Product Code. 10520591
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1530-32-1
Molecular Formula C20H20BrP
Molecular Weight (g/mol) 371.26
MDL Number MFCD00011838
InChI Key JHYNXXDQQHTCHJ-UHFFFAOYSA-M
Synonym ethyltriphenylphosphonium bromide, ethyl triphenyl phosphonium bromide, triphenylethylphosphonium bromide, phosphonium, ethyltriphenyl-, bromide, ethyltriphenylphosphanium bromide, ethyl triphenylphosphonium bromide, unii-r85v84ul5v, ethyltriphenylphosphoniumbromide, guaifenesin powder, pubchem18705
PubChem CID 73727
IUPAC Name ethyl(triphenyl)phosphanium;bromide
SMILES [Br-].CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 206.5°C to 208.5°C
Color White
Flash Point >200°C
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (on dry substance) (Argentometry)
Packaging Glass bottle
Linear Formula C2H5P(C6H5)3Br
Quantity 25 g
Solubility Information Solubility in water: 120g/L (23°C). Other solubilities: soluble in chloroform,fairly soluble in lower alcohols,slightly soluble in arom. hydrocarbons
Formula Weight 371.26
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Ethyltriphenylphosphonium bromide, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyltriphenylphosphonium bromide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'