missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress Ethynylcytidine
MedChemExpress Ethynylcytidine (ECyD), a nucleoside analog and a potent inhibitor of RNA synthesis, inhibits RNA polymerases I, II and II. Ethynylcytidine has robust antitumor activity in a wide range of models of cancer.
Brand: MedChemExpress HY-16200
This item is not returnable.
View return policy
Specifications
267.24 g/mol | |
Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month | |
98.96% | |
DMSO : 250 mg/mL (935.49 mM∣Need ultrasonic) | |
180300-43-0 |
Reference compound | |
Room temperature | |
25 mg | |
Cancer-programmed cell death | |
ECyD∣ TAS-106∣ 3'-C-Ethynylcytidine |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction