missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Etomidate, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
General anesthetic with GABA modulatory and GABA-mimetic actions; selectively interacts with β2- and β3-subunit containing GABAA receptors. Short acting and potent hypnotic, with low toxicity.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 244.29 |
Percent Purity | >99% |
Chemical Name or Material | Etomidate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'